Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 27 - 28 | 5518 | 70 | |
| GenBank mRNA | AK051474.1 | - | 4 - 5 | 746 | 70 | |
| AK122351.1 | - | 24 - 25 | 4508 | 70 | ||
| AK147675.1 | - | 25 - 26 | 6108 | 70 | ||
| AK162968.1 | - | 6 - 7 | 978 | 70 | ||
More Information
Set Membership: |
|

Cul9
Genomic Map