Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 28 - 29 | 5651 | 62 | |
| GenBank mRNA | AK051474.1 | - | 5 - 6 | 879 | 62 | |
| AK122351.1 | - | 25 - 26 | 4641 | 62 | ||
| AK147675.1 | - | 26 - 27 | 6241 | 62 | ||
| AK162968.1 | - | 7 - 8 | 1111 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map