Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 30 - 31 | 6105 | 78 | |
| GenBank mRNA | AK051474.1 | - | 7 - 8 | 1333 | 78 | |
| AK122351.1 | - | 27 - 28 | 5095 | 78 | ||
| AK147675.1 | - | 28 - 29 | 6695 | 78 | ||
| AK162968.1 | - | 9 - 10 | 1565 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map