Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 32 - 33 | 6592 | 96 | |
| GenBank mRNA | AK051474.1 | - | 9 - 10 | 1820 | 96 | |
| AK122351.1 | - | 29 - 30 | 5582 | 96 | ||
| AK147675.1 | - | 30 - 31 | 7182 | 96 | ||
| AK162968.1 | - | 11 - 12 | 2053 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map