Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 38 - 39 | 7315 | 84 | |
| GenBank mRNA | AK051474.1 | - | 15 - 16 | 2544 | 84 | |
| AK122351.1 | - | 35 - 36 | 6305 | 84 | ||
| AK147675.1 | - | 36 - 37 | 7929 | 84 | ||
More Information
Set Membership: |
|

Cul9
Genomic Map