Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 39 - 40 | 7451 | 77 | |
| GenBank mRNA | AK051474.1 | - | 16 - 17 | 2680 | 77 | |
| AK122351.1 | - | 36 - 37 | 6441 | 77 | ||
| AK147675.1 | - | 37 - 38 | 8065 | 77 | ||
| AK162968.1 | - | 18 - 19 | 2913 | 77 | ||
| BP753198.1 | - | 2 - 3 | 77 | |||
More Information
Set Membership: |
|

Cul9
Genomic Map