Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 10 - 11 | 2746 | 57 | |
| GenBank mRNA | AK122351.1 | - | 7 - 8 | 1737 | 57 | |
| AK147675.1 | - | 9 - 10 | 3001 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map