Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 12 - 13 | 3210 | 70 | |
| GenBank mRNA | AK122351.1 | - | 9 - 10 | 2201 | 70 | |
| AK147675.1 | - | 11 - 12 | 3465 | 70 | ||
More Information
Set Membership: |
|

Cul9
Genomic Map