Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 13 - 14 | 3310 | 76 | |
| GenBank mRNA | AK122351.1 | - | 10 - 11 | 2301 | 76 | |
| AK147675.1 | - | 12 - 13 | 3565 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map