Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 10 - 11 | 1361 | 62 | |
| NM_021359.3 | NP_067334.1 | 7 - 8 | 1255 | 62 | ||
| GenBank mRNA | AF115376.1 | - | 10 - 11 | 1207 | 62 | |
| AK008867.1 | - | 7 - 8 | 1254 | 62 | ||
| AK036439.1 | - | 10 - 11 | 1362 | 62 | ||
| AK085900.1 | - | 10 - 11 | 1402 | 62 | ||
| BC049185.1 | - | 7 - 8 | 1224 | 6475423 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb6
Genomic Map