Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 15 - 16 | 2314 | 77 | |
| NM_021359.3 | NP_067334.1 | 12 - 13 | 2208 | 77 | ||
| GenBank mRNA | AF115376.1 | - | 15 - 16 | 2160 | 77 | |
| AK036439.1 | - | 15 - 16 | 2315 | 77 | ||
| AK085900.1 | - | 15 - 16 | 2355 | 77 | ||
| BC049185.1 | - | 12 - 13 | 2177 | 6475423 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb6
Genomic Map