Assay Details
| Gene Symbol: Aars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198608.2 | NP_941010.2 | 11 - 12 | 1577 | 70 | |
| GenBank mRNA | AK146958.1 | - | 11 - 12 | 1577 | 70 | |
| AK167771.1 | - | 11 - 12 | 1572 | 70 | ||
| AK173139.1 | - | 7 - 8 | 843 | 70 | ||
| BC059889.1 | - | 2 - 3 | 606 | 70 | ||
| BC079844.1 | - | 11 - 12 | 1549 | 70 | ||
| BC110699.1 | - | 2 - 3 | 606 | 70 | ||
| BC113172.1 | - | 11 - 12 | 1573 | 40060953 | 70 | |
| BC113779.1 | - | 11 - 12 | 1573 | 40060952 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aars2
Genomic Map