Assay Details
| Gene Symbol: Aars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198608.2 | NP_941010.2 | 5 - 6 | 890 | 78 | |
| GenBank mRNA | AK146958.1 | - | 5 - 6 | 890 | 78 | |
| AK173139.1 | - | 1 - 2 | 156 | 78 | ||
| BC079844.1 | - | 5 - 6 | 862 | 78 | ||
| BC113172.1 | - | 5 - 6 | 886 | 40060953 | 78 | |
| BC113779.1 | - | 5 - 6 | 886 | 40060952 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aars2
Genomic Map