Assay Details
| Gene Symbol: Palb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081238.2 | NP_001074707.1 | 8 - 9 | 2765 | 71 | |
| NM_001289842.1 | NP_001276771.1 | 8 - 9 | 2765 | 71 | ||
| NM_001289843.1 | NP_001276772.1 | 7 - 8 | 2671 | 71 | ||
| NM_001289844.1 | NP_001276773.1 | 8 - 9 | 1676 | 71 | ||
| NM_001289845.1 | NP_001276774.1 | 5 - 6 | 605 | 71 | ||
| GenBank mRNA | AK154227.1 | - | 8 - 9 | 2737 | 71 | |
| AK156701.1 | - | 8 - 9 | 2684 | 71 | ||
| AK157382.1 | - | 8 - 9 | 2684 | 71 | ||
| BC044748.2 | - | 4 - 5 | 370 | 71 | ||
| BC055302.1 | - | 5 - 6 | 568 | 71 | ||
| BC066139.1 | - | 7 - 8 | 2613 | 71 | ||
| BC066140.1 | - | 8 - 9 | 1574 | 30459312 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Palb2
Genomic Map