Assay Details
| Gene Symbol: Palb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081238.2 | NP_001074707.1 | 11 - 12 | 3136 | 64 | |
| NM_001289844.1 | NP_001276773.1 | 11 - 12 | 2047 | 64 | ||
| NM_001289845.1 | NP_001276774.1 | 8 - 9 | 976 | 64 | ||
| GenBank mRNA | AK156701.1 | - | 11 - 12 | 3055 | 64 | |
| AK157382.1 | - | 11 - 12 | 3055 | 64 | ||
| BC044748.2 | - | 7 - 8 | 741 | 64 | ||
| BC055302.1 | - | 8 - 9 | 939 | 64 | ||
| BC066140.1 | - | 12 - 13 | 1945 | 30459312 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Palb2
Genomic Map