Assay Details
| Gene Symbol: Ate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029895.3 | NP_001025066.1 | 3 - 4 | 621 | 122 | |
| NM_001136054.2 | NP_001129526.1 | 3 - 4 | 353 | 122 | ||
| NM_001271343.1 | NP_001258272.1 | 3 - 4 | 621 | 122 | ||
| NM_013799.3 | NP_038827.2 | 3 - 4 | 353 | 122 | ||
| GenBank mRNA | AK138277.1 | - | 3 - 4 | 272 | 122 | |
| AK148442.1 | - | 3 - 4 | 321 | 122 | ||
| AK158613.1 | - | 3 - 4 | 623 | 122 | ||
| AK167591.1 | - | 3 - 4 | 313 | 122 | ||
| BC042601.1 | - | 3 - 4 | 312 | 4503326 | 122 | |
| BC050948.1 | - | 3 - 4 | 303 | 6826691 | 122 | |
| DQ093639.1 | - | 3 - 4 | 213 | 122 | ||
| DQ093640.1 | - | 3 - 4 | 213 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ate1
Genomic Map