Assay Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004468.2 | NP_001004468.1 | 14 - 15 | 7829 | 82 | |
| NM_021314.4 | NP_067289.2 | 10 - 11 | 2664 | 82 | ||
| NM_206856.3 | NP_996738.2 | 13 - 14 | 2630 | 82 | ||
| GenBank mRNA | AB041546.1 | - | 10 - 11 | 2665 | 82 | |
| AY177410.1 | - | 12 - 13 | 2642 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tacc2
Genomic Map