Assay Details
| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122640.1 | NP_001116112.1 | 9 - 10 | 838 | 91 | |
| NM_001122641.1 | NP_001116113.1 | 9 - 10 | 838 | 91 | ||
| NM_001122642.1 | NP_001116114.1 | 9 - 10 | 838 | 91 | ||
| NM_001122643.1 | NP_001116115.1 | 9 - 10 | 838 | 91 | ||
| NM_144529.2 | NP_653112.2 | 9 - 10 | 838 | 91 | ||
| GenBank mRNA | AK036468.1 | - | 9 - 10 | 775 | 91 | |
| AK036617.1 | - | 9 - 10 | 775 | 91 | ||
| AK147010.1 | - | 9 - 10 | 809 | 91 | ||
| AK149986.1 | - | 9 - 10 | 810 | 91 | ||
| AK152266.1 | - | 9 - 10 | 775 | 91 | ||
| AK152301.1 | - | 9 - 10 | 802 | 91 | ||
| AK161636.1 | - | 9 - 10 | 839 | 91 | ||
| BC003259.1 | - | 9 - 10 | 786 | 3500822 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap17
Genomic Map