Assay Details
| Gene Symbol: Zfp804a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175513.3 | NP_780722.2 | 3 - 4 | 518 | 78 | |
| GenBank mRNA | AK043989.1 | - | 2 - 3 | 911 | 78 | |
| AK049881.1 | - | 3 - 4 | 519 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp804a
Genomic Map