Assay Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178115.4 | NP_835216.3 | 4 - 5 | 652 | 106 | |
| GenBank mRNA | AK052387.1 | - | 4 - 5 | 635 | 106 | |
| AK144179.1 | - | 4 - 5 | 613 | 106 | ||
| AK144959.1 | - | 4 - 5 | 623 | 106 | ||
| BC072596.1 | - | 4 - 5 | 621 | 6409214 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Edrf1
Genomic Map