Assay Details
| Gene Symbol: Nt5c1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027588.3 | NP_081864.2 | 3 - 4 | 364 | 73 | |
| GenBank mRNA | AJ295253.1 | - | 3 - 4 | 291 | 73 | |
| AK014892.1 | - | 3 - 4 | 364 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c1b
Genomic Map