Assay Details
| Gene Symbol: Nt5c1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356186.1 | - | 4 - 5 | 801 | 66 | |
| BC060976.1 | - | 4 - 5 | 820 | 6773248 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c1b
Genomic Map