Assay Details
| Gene Symbol: Nt5c1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027588.3 | NP_081864.2 | 7 - 8 | 1297 | 118 | |
| GenBank mRNA | AF356186.1 | - | 7 - 8 | 1216 | 118 | |
| AJ295253.1 | - | 7 - 8 | 1224 | 118 | ||
| AK014892.1 | - | 7 - 8 | 1297 | 118 | ||
| BC060976.1 | - | 7 - 8 | 1235 | 6773248 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c1b
Genomic Map