Assay Details
| Gene Symbol: Ppp2r2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026391.2 | NP_080667.1 | 1 - 2 | 128 | 108 | |
| GenBank mRNA | AK011198.1 | - | 1 - 2 | 150 | 108 | |
| AK129386.2 | - | 1 - 2 | 155 | 108 | ||
| BC066022.1 | - | 1 - 2 | 128 | 6828707 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066022.1, NM_026391.2 transcripts.
Set Membership: |
|

Ppp2r2d
Genomic Map