Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 10 - 11 | 1605 | 85 | |
| GenBank mRNA | AF171092.1 | - | 10 - 11 | 1610 | 85 | |
| AK137092.1 | - | 10 - 11 | 1505 | 85 | ||
| BC058675.1 | - | 10 - 11 | 1417 | 6834418 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map