Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 19 - 20 | 2633 | 115 | |
| GenBank mRNA | AF171092.1 | - | 19 - 20 | 2638 | 115 | |
| AK137092.1 | - | 19 - 20 | 2533 | 115 | ||
| AK162228.1 | - | 2 - 3 | 135 | 115 | ||
| BC058675.1 | - | 19 - 20 | 2445 | 6834418 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map