Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 25 - 26 | 3460 | 73 | |
| GenBank mRNA | AF171092.1 | - | 25 - 26 | 3465 | 73 | |
| AK078018.1 | - | 2 - 3 | 226 | 73 | ||
| AK137092.1 | - | 25 - 26 | 3360 | 73 | ||
| AK162228.1 | - | 8 - 9 | 962 | 73 | ||
| BC058675.1 | - | 25 - 26 | 3272 | 6834418 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map