Assay Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146119.1 | NP_001139591.1 | 8 - 9 | 883 | 65 | |
| NM_029758.4 | NP_084034.2 | 9 - 10 | 956 | 65 | ||
| GenBank mRNA | AK010821.1 | - | 7 - 8 | 819 | 65 | |
| AK047142.1 | - | 8 - 9 | 846 | 65 | ||
| BC049664.1 | - | 9 - 10 | 918 | 6532813 | 65 | |
| BC052465.1 | - | 8 - 9 | 864 | 6834768 | 65 | |
More Information
Set Membership: |
|

Fam49a
Genomic Map