Assay Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008434.2 | NP_032460.2 | 14 - 15 | 1830 | 58 | |
| GenBank mRNA | AK136389.1 | - | 7 - 8 | 700 | 58 | |
| AK154244.1 | - | 14 - 15 | 1804 | 58 | ||
| AY331142.1 | - | 14 - 15 | 1745 | 58 | ||
| BC045142.1 | - | 14 - 15 | 1830 | 6314642 | 58 | |
| BC055304.1 | - | 14 - 15 | 1785 | 6397978 | 58 | |
| U70068.1 | - | 14 - 15 | 1636 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq1
Genomic Map