Assay Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008434.2 | NP_032460.2 | 7 - 8 | 1131 | 59 | |
| GenBank mRNA | AB079603.1 | - | 7 - 8 | 834 | 59 | |
| AK154244.1 | - | 7 - 8 | 1105 | 59 | ||
| AY331142.1 | - | 7 - 8 | 1046 | 59 | ||
| AY644390.1 | - | 2 - 3 | 239 | 59 | ||
| BC045142.1 | - | 7 - 8 | 1131 | 6314642 | 59 | |
| BC055304.1 | - | 7 - 8 | 1086 | 6397978 | 59 | |
| U70068.1 | - | 7 - 8 | 937 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq1
Genomic Map