Assay Details
| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011859.3 | NP_035989.1 | 1 - 2 | 294 | 76 | |
| GenBank mRNA | BC023922.1 | - | 1 - 2 | 250 | 5323948 | 76 |
| BQ832175.1 | - | 1 - 2 | 318 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023922.1, NM_011859.3 transcripts.
Set Membership: |
|

Osr1
Genomic Map