Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_001130412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_001130412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_015763.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_015763.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_172950.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
NM_172950.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010195.mm.1 |
TC1200001460.mm.1 |
MTA TC |