Assay Details
| Gene Symbol: Scn2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099298.2 | NP_001092768.1 | 11 - 12 | 2071 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn2a1| Gene Symbol: Scn2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099298.2 | NP_001092768.1 | 11 - 12 | 2071 | 84 | |
Set Membership: |
|