Assay Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172671.2 | NP_766259.2 | 12 - 13 | 1585 | 97 | |
| GenBank mRNA | AK044357.1 | - | 11 - 12 | 893 | 97 | |
| AK052660.1 | - | 1 - 2 | 594 | 97 | ||
| CN667965.1 | - | 3 - 4 | 157 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgr4
Genomic Map