Assay Details
| Gene Symbol: Cdc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152810.2 | NP_690023.1 | 7 - 8 | 1175 | 99 | |
| GenBank mRNA | AK004547.2 | - | 7 - 8 | 1205 | 99 | |
| AK146892.1 | - | 7 - 8 | 995 | 99 | ||
| AK150562.1 | - | 7 - 8 | 992 | 99 | ||
| AK150861.1 | - | 7 - 8 | 992 | 99 | ||
| AK172950.1 | - | 7 - 8 | 995 | 99 | ||
| BC003893.1 | - | 7 - 8 | 1000 | 3593535 | 99 | |
| BC031480.1 | - | 7 - 8 | 1175 | 3498853 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdc5l
Genomic Map