Assay Details
| Gene Symbol: Spi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011355.1 | NP_035485.1 | 2 - 3 | 291 | 59 | |
| GenBank mRNA | AK142246.1 | - | 2 - 3 | 308 | 59 | |
| AK153524.1 | - | 2 - 3 | 322 | 59 | ||
| AK154945.1 | - | 2 - 3 | 343 | 59 | ||
| AK155085.1 | - | 2 - 3 | 327 | 59 | ||
| BC003815.1 | - | 2 - 3 | 209 | 3600260 | 59 | |
| L03215.1 | - | 2 - 3 | 188 | 59 | ||
| M32370.1 | - | 2 - 3 | 271 | 59 | ||
| M38252.1 | - | 2 - 3 | 291 | 59 | ||
| X17463.1 | - | 2 - 3 | 979 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spi1
Genomic Map