Assay Details
| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 4 - 5 | 719 | 124 | |
| GenBank mRNA | AF099988.1 | - | 4 - 5 | 695 | 124 | |
| AK017292.1 | - | 4 - 5 | 677 | 124 | ||
| AK148456.1 | - | 4 - 5 | 756 | 124 | ||
| BC051964.1 | - | 4 - 5 | 1006 | 5698326 | 124 | |
| BC064443.1 | - | 4 - 5 | 719 | 6843981 | 124 | |
| EF410085.1 | - | 512 | 124 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Stk39
Genomic Map