Assay Details
| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 6 - 7 | 875 | 63 | |
| GenBank mRNA | AF099988.1 | - | 6 - 7 | 851 | 63 | |
| AK148456.1 | - | 6 - 7 | 912 | 63 | ||
| BC051964.1 | - | 6 - 7 | 1162 | 5698326 | 63 | |
| BC064443.1 | - | 6 - 7 | 875 | 6843981 | 63 | |
| EF410085.1 | - | 668 | 63 | |||
| JN368425.1 | - | 2 - 3 | 456 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the JN368425.1 transcripts.
Set Membership: |
|

Stk39
Genomic Map