Assay Details
| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016866.2 | NP_058562.1 | 7 - 8 | 978 | 64 | |
| GenBank mRNA | AF099988.1 | - | 7 - 8 | 954 | 64 | |
| AK017292.1 | - | 7 - 8 | 938 | 64 | ||
| AK148456.1 | - | 7 - 8 | 1015 | 64 | ||
| BC051964.1 | - | 7 - 8 | 1265 | 5698326 | 64 | |
| BC064443.1 | - | 7 - 8 | 978 | 6843981 | 64 | |
| EF410085.1 | - | 771 | 64 | |||
| JN368425.1 | - | 3 - 4 | 559 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk39
Genomic Map