Assay Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031811.2 | NP_001026981.2 | 11 - 12 | 2136 | 79 | |
| GenBank mRNA | AK048629.1 | - | 11 - 12 | 2025 | 79 | |
| BC029690.1 | - | 7 - 8 | 1363 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh8
Genomic Map