Assay Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031811.2 | NP_001026981.2 | 13 - 14 | 2523 | 101 | |
| GenBank mRNA | AK048629.1 | - | 13 - 14 | 2412 | 101 | |
| BC029690.1 | - | 9 - 10 | 1750 | 101 | ||
| BC130020.1 | - | 2 - 3 | 661 | 101 | ||
| BC152323.1 | - | 2 - 3 | 661 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC130020.1 transcripts.
Set Membership: |
|

Kcnh8
Genomic Map