Assay Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031811.2 | NP_001026981.2 | 3 - 4 | 539 | 97 | |
| GenBank mRNA | AK048629.1 | - | 3 - 4 | 428 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh8
Genomic Map