Assay Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031811.2 | NP_001026981.2 | 5 - 6 | 894 | 89 | |
| GenBank mRNA | AK048629.1 | - | 5 - 6 | 783 | 89 | |
| BC029690.1 | - | 1 - 2 | 121 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh8
Genomic Map