Assay Details
| Gene Symbol: Kcnh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031811.2 | NP_001026981.2 | 6 - 7 | 1070 | 68 | |
| GenBank mRNA | AK048629.1 | - | 6 - 7 | 959 | 68 | |
| BC029690.1 | - | 2 - 3 | 297 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh8
Genomic Map