Assay Details
| Gene Symbol: Cstf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145529.3 | NP_663504.1 | 14 - 15 | 1460 | 145 | |
| GenBank mRNA | AK159180.1 | - | 14 - 15 | 1263 | 145 | |
| BC003241.1 | - | 14 - 15 | 1421 | 3498424 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cstf3
Genomic Map