Assay Details
| Gene Symbol: Cers6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172856.3 | NP_766444.1 | 7 - 8 | 820 | 88 | |
| GenBank mRNA | AK028849.1 | - | 7 - 8 | 821 | 88 | |
| AK135798.1 | - | 7 - 8 | 922 | 88 | ||
| AK151848.1 | - | 7 - 8 | 909 | 88 | ||
| AK152417.1 | - | 7 - 8 | 890 | 88 | ||
| BC057629.1 | - | 7 - 8 | 863 | 4224547 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers6
Genomic Map