Assay Details
| Gene Symbol: Xpo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028816.2 | NP_083092.2 | 5 - 6 | 1069 | 80 | |
| GenBank mRNA | AK049009.1 | - | 5 - 6 | 1082 | 80 | |
| AK154709.1 | - | 5 - 6 | 1111 | 80 | ||
| BC058090.1 | - | 5 - 6 | 1069 | 6826356 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpo6
Genomic Map