Assay Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163818.1 | NP_001157290.1 | 3 - 4 | 719 | 81 | |
| GenBank mRNA | AJ504718.1 | - | 3 - 4 | 725 | 81 | |
| AK053183.1 | - | 4 - 5 | 738 | 81 | ||
| AK145485.1 | - | 2 - 3 | 163 | 81 | ||
| BC030903.1 | - | 3 - 4 | 726 | 81 | ||
| BC052059.1 | - | 3 - 4 | 722 | 5702171 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppp1r10
Genomic Map