Assay Details
| Gene Symbol: Pla2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145378.4 | NP_663353.3 | 8 - 9 | 709 | 57 | |
| GenBank mRNA | AK037218.1 | - | 6 - 7 | 1931 | 57 | |
| AK137721.1 | - | 8 - 9 | 710 | 57 | ||
| DQ888308.1 | - | 7 - 8 | 617 | 57 | ||
| Gene Symbol: Gm28042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104353.1 | 15 - 16 | 1952 | 57 | ||
| GenBank mRNA | AB195274.1 | - | 15 - 16 | 1942 | 57 | |
| AB195275.1 | - | 14 - 15 | 4050 | 57 | ||
| BC098210.1 | - | 9 - 10 | 1103 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Pla2g4b
Genomic Map