Assay Details
| Gene Symbol: Pla2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145378.4 | NP_663353.3 | 16 - 17 | 1683 | 68 | |
| GenBank mRNA | AK037218.1 | - | 13 - 14 | 3132 | 68 | |
| AK137721.1 | - | 15 - 16 | 1912 | 68 | ||
| BC042758.1 | - | 4 - 5 | 562 | 68 | ||
| DQ888308.1 | - | 15 - 16 | 1591 | 68 | ||
| Gene Symbol: Gm28042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104353.1 | 23 - 24 | 2926 | 68 | ||
| GenBank mRNA | AB195274.1 | - | 23 - 24 | 2916 | 68 | |
| AB195275.1 | - | 22 - 23 | 5024 | 68 | ||
| BC098210.1 | - | 17 - 18 | 2077 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Pla2g4b
Genomic Map